Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics Q-Slide Gram, Pre-inoculated with E. coli and S. aureus
SDP

Catalog No. 22026379 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Target Organism Class:
Bacteria, Fungi, Yeast, Molds
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Organism Class
22-026-379 Bacteria, Fungi, Yeast, Molds
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-026-379 Supplier Hardy Diagnostics Supplier No. Z302
Add to Cart
Add to Cart

A convenient ready-to-stain control slide that will test both your stain solutions and your staining techniques.

  • Positive and negative stain control organisms are affixed to the circles in each Q-Slide™
  • Up to four patient specimens (in the squares) may be processed at the same time as the QC organisms
  • AFB - for your Acid Fast Bacillus stain QC; either Kinyoun or Ziehl Neelson
  • Contains Mycobacterium gordonae as a positive control and a corynebacterium as a negative control
  • Gram Stain QC contains colias a Gram-negative control, and a staphylococcus aureus as a Gram-positive control
  • Quick and effective way to ensure the quality of reagents used as well as the proficiency of staining technique by laboratory personnel

Specifications

Manufacturing Origin United States
Product Type Contact Slide
Sterility Non-sterile
Target Organism Class Bacteria, Fungi, Yeast, Molds
Test Type Microbial Detection, Microbial Identification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.