Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Q-Tof Qualification Standards Kit, 11 Components

Catalog No. 50201677 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Mass Spectrometry (QToF) standards kit is designed for system assessment and contains 11 components for qualification purposes with Qtof MS instruments.

  • Designed for system assessment in mass spectrometry (QToF).
  • Includes a certificate of analysis.
  • Storage condition: –30°C to –10°C (Freezer).
  • Shelf life: 6 months.
  • Shipping condition: Ambient.
  • Supports reversed phase separation mode.
  • Utilizes MS detection method.
  • Available in liquid and solid formulations.
  • Compatible with Qtof MS system types.
  • Intended for system performance application.
  • One unit per package.

Catalog No. 50-201-677 Supplier Waters Corp Supplier No. 700003276
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Mass Spectrometry (QToF) standards kit is designed for system assessment and contains 11 components for qualification purposes with Qtof MS instruments.

  • Designed for system assessment in mass spectrometry (QToF).
  • Includes a certificate of analysis.
  • Storage condition: –30°C to –10°C (Freezer).
  • Shelf life: 6 months.
  • Shipping condition: Ambient.
  • Supports reversed phase separation mode.
  • Utilizes MS detection method.
  • Available in liquid and solid formulations.
  • Compatible with Qtof MS system types.
  • Intended for system performance application.
  • One unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.