Learn More
Sigma Aldrich Fine Chemicals Biosciences Q TubeTM gas purging set 11EA
Catalog No. 502908790
Encompass
The parts with direct chemical contact are made of 316 stainless steel PTFE Simriz or glass.Q-Tube-Purging-12-SS / 100-QT00PG-012SS can be operated safely and accurately with high reproducibility with a wide range of gases including HCl NH3 H2 O2 N2 Ar D2 CO CO2 and etc. The set contains all items needed for immediate use.
Catalog No. 50-290-8790 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7424191EA
The parts with direct chemical contact are made of 316 stainless steel PTFE Simriz or glass.Q-Tube-Purging-12-SS / 100-QT00PG-012SS can be operated safely and accurately with high reproducibility with a wide range of gases including HCl NH3 H2 O2 N2 Ar D2 CO CO2 and etc. The set contains all items needed for immediate use.