Learn More
Sigma Aldrich Fine Chemicals Biosciences Q TubeTM gas purging set p1EA
Catalog No. 502908563
Encompass
This set can be operated safely and accurately with high reproducibility with a wide range of gases including oxygen nitrogen argon hydrogen deuterium carbon monoxide and carbon dioxide.The set contains all items needed for immediate use316 stainless steel adapter and matched PTFE sleeve1 pressure gauge 300 psi 2 valves1 pressure tube 35 mL20 septaAll fittings for connecting to a gas regulator
Catalog No. 50-290-8563 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7579931EA
This set can be operated safely and accurately with high reproducibility with a wide range of gases including oxygen nitrogen argon hydrogen deuterium carbon monoxide and carbon dioxide.The set contains all items needed for immediate use316 stainless steel adapter and matched PTFE sleeve1 pressure gauge 300 psi 2 valves1 pressure tube 35 mL20 septaAll fittings for connecting to a gas regulator