Learn More
Sigma Aldrich Fine Chemicals Biosciences Q TubeTM pressure tube rea1EA
Catalog No. 502909027
Encompass
Q-Tube pressure reactors are affordable alternatives to a microwave synthesizer and feature a safe pressure-release system that prevents accidental explosions due to over -pressurization. The starter kits contain all items needed for immediate use.Optional higher pressure-release threshold adapters are available separately for 12 mL and 35 mL Q-Tubes.BenefitsBetter yield than microwave and conventional open flaskCleaner product than microwave and conventional open flask (higher yield and lighter color)Reduced reaction time (from hours to minutes)Higher reproducibility than microwaveScalable (up to 20 grams)Safer (automatic pressure release and reseal) than conventional pressure (sealed) tubeAffordableMaintenance-free
Catalog No. 50-290-9027 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5676711EA
Q-Tube pressure reactors are affordable alternatives to a microwave synthesizer and feature a safe pressure-release system that prevents accidental explosions due to over -pressurization. The starter kits contain all items needed for immediate use.Optional higher pressure-release threshold adapters are available separately for 12 mL and 35 mL Q-Tubes.BenefitsBetter yield than microwave and conventional open flaskCleaner product than microwave and conventional open flask (higher yield and lighter color)Reduced reaction time (from hours to minutes)Higher reproducibility than microwaveScalable (up to 20 grams)Safer (automatic pressure release and reseal) than conventional pressure (sealed) tubeAffordableMaintenance-free