Learn More
Medchemexpress LLC HY-12305 10mg Medchemexpress, Q-VD-OPh CAS:1135695-98-5 Purity:>98%
SDP
Catalog No. 502028872
Encompass_Preferred
Medchemexpress, HY-12305 10mg Q-VD-OPh CAS:1135695-98-5 Q-VD-OPh is an irreversible pan-caspase inhibitor with potent antiapoptotic properties; inhibits caspase 7 with an IC50 of 48 nM and 25-400 nM for other caspases including caspase 1, 3, 8, 9, 10, and 12. Q-VD-OPh can inhibits HIV infection. Q-VD-OPh is able to cross the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8872 Supplier Medchemexpress LLC Supplier No. HY1230510MG
Medchemexpress, HY-12305 10mg Q-VD-OPh CAS:1135695-98-5 Q-VD-OPh is an irreversible pan-caspase inhibitor with potent antiapoptotic properties; inhibits caspase 7 with an IC50 of 48 nM and 25-400 nM for other caspases including caspase 1, 3, 8, 9, 10, and 12. Q-VD-OPh can inhibits HIV infection. Q-VD-OPh is able to cross the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.