Learn More
Medchemexpress LLC Q-VD-OPh | 1135695-98-5 | MFCD08669741 | 99.9% | 513.49 g/mol | C26H25F2N3O6 | 10MM 1ML
SDP
Catalog No. 5000235743
Encompass_Preferred
Q-VD-OPh is an irreversible pan-caspase inhibitor used in research to block apoptosis. It inhibits caspase-7 with an IC50 of approximately 48 nM and shows inhibitory activity against multiple caspases (IC50 25-400 nM). The compound is cell-permeable, reported to cross the blood-brain barrier, and is provided as powder and as DMSO stock solutions for convenient preparation of working concentrations.
  • Irreversible pan-caspase inhibitor with low-nanomolar potency against caspase-7.
  • Activity against multiple caspases (IC50 range 25-400 nM).
  • Cell-permeable and reported to cross the blood-brain barrier.
  • Available as powder or pre-prepared DMSO stock for flexible use.
  • High purity suitable for research applications (reported 99.9%).
Catalog No. 50-002-35743 Supplier Medchemexpress LLC Supplier No. HY1230510MM 1ML
Q-VD-OPh is an irreversible pan-caspase inhibitor used in research to block apoptosis. It inhibits caspase-7 with an IC50 of approximately 48 nM and shows inhibitory activity against multiple caspases (IC50 25-400 nM). The compound is cell-permeable, reported to cross the blood-brain barrier, and is provided as powder and as DMSO stock solutions for convenient preparation of working concentrations.
  • Irreversible pan-caspase inhibitor with low-nanomolar potency against caspase-7.
  • Activity against multiple caspases (IC50 range 25-400 nM).
  • Cell-permeable and reported to cross the blood-brain barrier.
  • Available as powder or pre-prepared DMSO stock for flexible use.
  • High purity suitable for research applications (reported 99.9%).