Learn More
Selleck Chemical LLC Q-VD-Oph 1mg 1135695-98-5
SDP
Supplier:  Selleck Chemical LLC S73111MG
Encompass_Preferred
Q-VD-Oph is a potent pan-caspase inhibitor with IC50 ranged from 25 to 400 nM for caspases 1,3,8, and 9. Q-VD-OPh can inhibits HIV infection. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4630