Learn More
Apexbio Technology LLC Q-VD-OPh 1135695-98-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A19015.1
Encompass_Preferred
Q-VD-OPh is a pan-caspase inhibitor targeting caspases including 1 3 8 and 9 with IC50 values ranging from approximately 25 to 400 nM By blocking multiple caspase-mediated apoptotic pathways including caspase-9/3 caspase-8/10 and caspase-12 pathways Q-VD-OPh prevents apoptotic cell death induced by agents such as actinomycin D In research contexts this inhibitor serves as a tool to investigate apoptosis mechanisms across diverse cell types and species including human mouse and rat It is also used to enhance cell viability upon thawing from cryopreservation under standard cryoprotectant conditions
Catalog No. 50-000-63708