Learn More
AdipoGen Q-VD-OPh
SDP
Catalog No. 502562459
Encompass_Preferred
Chemical. CAS 1135695-98-5 anhydrous. Formula C26H25F2N3O6. MW 513.5. Synthetic. Cell permeable, irreversible and non-toxic non-FMK pan-caspase inhibitor with improved potency, stability and toxicity over Z-VAD-FMK. Does not cross-react with cathepsins nor calpains. Non-toxic due to the 2,6-difluorophenoxy methyl OPh group. The peptide is not O-methylated to reduce hydrophobicity and to facilitate use in aqueous media. Inhibits ICE-family protease/caspase processing, leading to apoptosis and autophagy induction. Decreases proteasome activity. Used in apoptosis and inflammasome studies.
Catalog No. 50-256-2459 Supplier AdipoGen Supplier No. AGCP30006M005
Chemical. CAS 1135695-98-5 anhydrous. Formula C26H25F2N3O6. MW 513.5. Synthetic. Cell permeable, irreversible and non-toxic non-FMK pan-caspase inhibitor with improved potency, stability and toxicity over Z-VAD-FMK. Does not cross-react with cathepsins nor calpains. Non-toxic due to the 2,6-difluorophenoxy methyl OPh group. The peptide is not O-methylated to reduce hydrophobicity and to facilitate use in aqueous media. Inhibits ICE-family protease/caspase processing, leading to apoptosis and autophagy induction. Decreases proteasome activity. Used in apoptosis and inflammasome studies.