Learn More
Medchemexpress LLC HY-12305 50mg , Q-VD-OPh QVD-OPH;Quinoline-Val-Asp-Difluorophenoxymethylketone CAS:1135695-98-5 Purity:98%
SDP
Catalog No. 501493637
Encompass_Preferred
Medchemexpress, HY-12305 50mg Q-VD-OPh QVD-OPH;Quinoline-Val-Asp-Difluorophenoxymethylketone CAS:1135695-98-5 Formula:C26H25F2N3O6 IC50: 48 nM (caspase 7), 25-400 nM (caspase 1, 3, 8, 9, 10, and 12) Purity:98% Q-VD-OPha is a irreversible pan-caspase inhibitor with potent antiapoptotic properties; inhibits caspase 7 with an IC 50 of 48 nM and 25-400 nM for other caspases including caspase 1, 3, 8, 9, 10, and 12. Q-VD-OPha is able to cross the blood-brain barrier. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3637 Supplier Medchemexpress LLC Supplier No. HY1230550MG
Medchemexpress, HY-12305 50mg Q-VD-OPh QVD-OPH;Quinoline-Val-Asp-Difluorophenoxymethylketone CAS:1135695-98-5 Formula:C26H25F2N3O6 IC50: 48 nM (caspase 7), 25-400 nM (caspase 1, 3, 8, 9, 10, and 12) Purity:98% Q-VD-OPha is a irreversible pan-caspase inhibitor with potent antiapoptotic properties; inhibits caspase 7 with an IC 50 of 48 nM and 25-400 nM for other caspases including caspase 1, 3, 8, 9, 10, and 12. Q-VD-OPha is able to cross the blood-brain barrier. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.