Learn More
Selleck Chemical LLC Q-VD-Oph
SDP
Catalog No. 5000004564
Encompass_Preferred
Q-VD-Oph (Quinoline-Val-Asp-Difluorophenoxymethylketone) is a potent pan-caspase inhibitor with IC50 ranged from 25 to 400 nM for caspases 1 3 8 and 9 Q-VD-OPh can inhibits HIV infection
Catalog No. 50-000-04564 Supplier Selleck Chemical LLC Supplier No. S7311100MG
Q-VD-Oph (Quinoline-Val-Asp-Difluorophenoxymethylketone) is a potent pan-caspase inhibitor with IC50 ranged from 25 to 400 nM for caspases 1 3 8 and 9 Q-VD-OPh can inhibits HIV infection