Learn More
Apexbio Technology LLC Q-VD(OMe)-OPh 10mg
SDP
Catalog No. 5000061642
Encompass_Preferred
Q-VD(OMe)-OPh is a pan-caspase inhibitor structurally identified as quinolyl-valyl-O-methylaspartyl-[2 6-difluorophenoxy]-methyl ketone As a broad-spectrum inhibitor Q-VD(OMe)-OPh reversibly targets multiple caspases involved in apoptotic signaling including initiator caspases (such as caspases 8 9 10 and 12) and executioner caspases (such as caspase 3) thereby blocking intrinsic extrinsic and endoplasmic reticulum-mediated apoptotic pathways In recombinant enzyme assays this inhibitor displays nanomolar-range IC values against caspases 1 3 8 and 9 Due to minimal cytotoxicity at tested concentrations Q-VD(OMe)-OPh serves as a tool compound in apoptosis-related biomedical research applied broadly for dissecting caspase-dependent apoptotic mechanisms and evaluating cytoprotective strategies
Catalog No. 50-000-61642 Supplier Apexbio Technology LLC Supplier No. A816510
Q-VD(OMe)-OPh is a pan-caspase inhibitor structurally identified as quinolyl-valyl-O-methylaspartyl-[2 6-difluorophenoxy]-methyl ketone As a broad-spectrum inhibitor Q-VD(OMe)-OPh reversibly targets multiple caspases involved in apoptotic signaling including initiator caspases (such as caspases 8 9 10 and 12) and executioner caspases (such as caspase 3) thereby blocking intrinsic extrinsic and endoplasmic reticulum-mediated apoptotic pathways In recombinant enzyme assays this inhibitor displays nanomolar-range IC values against caspases 1 3 8 and 9 Due to minimal cytotoxicity at tested concentrations Q-VD(OMe)-OPh serves as a tool compound in apoptosis-related biomedical research applied broadly for dissecting caspase-dependent apoptotic mechanisms and evaluating cytoprotective strategies