Learn More
AdipoGen Q-VE-OPh Neg Ctrl Q-VD-OPh
SDP
Catalog No. 502562458
Encompass_Preferred
Chemical. Formula C27H27F2N3O6. MW 527.5. Synthetic. Cell permeable, non-toxic negative control for the caspase inhibitor Q-VD-OPh Prod. No. AG-CP3-0006. Stable in aqueous environment. Several folds less inhibitory activity than Q-VD-OPh. Perfect for both in vitro and in vivo studies.
Catalog No. 50-256-2458 Supplier AdipoGen Supplier No. AGCP30007M001
Chemical. Formula C27H27F2N3O6. MW 527.5. Synthetic. Cell permeable, non-toxic negative control for the caspase inhibitor Q-VD-OPh Prod. No. AG-CP3-0006. Stable in aqueous environment. Several folds less inhibitory activity than Q-VD-OPh. Perfect for both in vitro and in vivo studies.