Learn More
Cayman Chemical Q1-ALC-0315 (iodide), Purity: ≥98%, CAS: 3112713-48-8, 10 mg
SDP
Catalog No. 502944979
Encompass_Preferred
A cationic quaternary ammonium lipid derivative of ALC-0315; LNPs containing Q1-ALC-0315 and ALC-0315 as a CLP and encapsulating mRNA encoding a luciferase reporter induce luciferase expression in the lungs and spleen in mice
Catalog No. 50-294-4979 Supplier Cayman Chemical Supplier No. 4124010 MG
A cationic quaternary ammonium lipid derivative of ALC-0315; LNPs containing Q1-ALC-0315 and ALC-0315 as a CLP and encapsulating mRNA encoding a luciferase reporter induce luciferase expression in the lungs and spleen in mice