Learn More
Cayman Chemical Q1-SM-102IodIde 10mg
SDP
Catalog No. 502944816
Encompass_Preferred
A cationic quaternary ammonium lipid derivative of SM-102; LNPs containing Q1-SM-102 and SM-102 as a CLP and encapsulating mRNA encoding a luciferase reporter induce luciferase expression in the lungs and spleen in mice
Catalog No. 50-294-4816 Supplier Cayman Chemical Supplier No. 4123910 MG
A cationic quaternary ammonium lipid derivative of SM-102; LNPs containing Q1-SM-102 and SM-102 as a CLP and encapsulating mRNA encoding a luciferase reporter induce luciferase expression in the lungs and spleen in mice