Learn More
Medchemexpress LLC HY-101040 5mg Medchemexpress, Q203 CAS:1334719-95-7 Purity:>98%
SDP
Catalog No. 501932019
Encompass_Preferred
Medchemexpress, HY-101040 5mg Q203 CAS:1334719-95-7 Q203 (IAP6) is a midazopyridine amide compound. Q203 is active against Mycobacterium tuberculosis H37Rv with an MIC 50 of 2.7 nM in culture broth medium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2019 Supplier Medchemexpress LLC Supplier No. HY1010405MG
Add to Cart
Add to Cart
Medchemexpress, HY-101040 5mg Q203 CAS:1334719-95-7 Q203 (IAP6) is a midazopyridine amide compound. Q203 is active against Mycobacterium tuberculosis H37Rv with an MIC 50 of 2.7 nM in culture broth medium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.