Learn More
Medchemexpress LLC HY-101040 100mg , Q203 IAP6;Telacebec CAS:1334719-95-7 Purity:98%
SDP
Catalog No. 501492979
Encompass_Preferred
Medchemexpress, HY-101040 100mg Q203 IAP6;Telacebec CAS:1334719-95-7 Formula:C29H28ClF3N4O2 MIC50: 2.7 nM ( Mycobacterium tuberculosis H37Rv) Purity:98% Q203 (IAP6) is a midazopyridine amide compound. Q203 is active against Mycobacterium tuberculosis H37Rv with an MIC 50 of 2.7 nM in culture broth medium. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2979 Supplier Medchemexpress LLC Supplier No. HY101040100MG
Medchemexpress, HY-101040 100mg Q203 IAP6;Telacebec CAS:1334719-95-7 Formula:C29H28ClF3N4O2 MIC50: 2.7 nM ( Mycobacterium tuberculosis H37Rv) Purity:98% Q203 (IAP6) is a midazopyridine amide compound. Q203 is active against Mycobacterium tuberculosis H37Rv with an MIC 50 of 2.7 nM in culture broth medium. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.