Learn More
Azzota Corp Q25 Semi-Micro 20mm blk Qz
Catalog No. 5000887078
Encompass_Preferred
Q25 Semi-Micro cell with black walls and lid 20mm Pathlength 4mm Inside Width Volume - 2 8ml Quartz
Catalog No. 50-008-87078 Supplier Azzota Corp Supplier No. Q25
Q25 Semi-Micro cell with black walls and lid 20mm Pathlength 4mm Inside Width Volume - 2 8ml Quartz