Learn More
New England Biolabs, Inc. Q5 High-Fidelity PCR Kit – 200 reactions
SDP
Catalog No. 50591064
Encompass
Q5 High-Fidelity DNA Polymerase sets a new standard for both fidelity and robust performance. With the highest fidelity amplification available (~280 times higher than Taq), Q5 DNA Polymerase results in ultra-low error rates. Q5 DNA Polymerase is composed of a novel polymerase that is fused to the processivity-enhancing Sso7d DNA binding domain, improving speed, fidelity and reliability of performance. This kit contains a sufficient supply of master mix and nuclease-free water to perform 50 polymerase chain reactions or 200 reactions. For added convenience, gel loading dye, and a Quick-Load DNA LAdder are also included in this kit.
Catalog No. 50-591-064 Supplier New England Biolabs, Inc. Supplier No. E0555L
Q5 High-Fidelity DNA Polymerase sets a new standard for both fidelity and robust performance. With the highest fidelity amplification available (~280 times higher than Taq), Q5 DNA Polymerase results in ultra-low error rates. Q5 DNA Polymerase is composed of a novel polymerase that is fused to the processivity-enhancing Sso7d DNA binding domain, improving speed, fidelity and reliability of performance. This kit contains a sufficient supply of master mix and nuclease-free water to perform 50 polymerase chain reactions or 200 reactions. For added convenience, gel loading dye, and a Quick-Load DNA LAdder are also included in this kit.