Learn More
New England Biolabs, Inc. Q5 Quick-Load™ Hot Start High-Fidelity 2X Master Mix - 500 reactions
SDP
Catalog No. 5000332609
Encompass
The Q5 Quick-Load Hot Start High-Fidelity 2X Master Mix offers high-fidelity performance in a convenient master mix format. The master mix features Q5, a high-fidelity, thermostable DNA polymerase with 3' to 5' exonuclease activity fused to a processivity-enhancing Sso7d domain. With an error rate ~280-fold lower than that of Taq DNA Polymerase, Q5 offers highly accurate amplification and can be used to amplify difficult or long targets (up to 20 kb for simple templates). The addition of a hot start aptamer inhibits polymerase activity at room temperature. The master mix includes hot start polymerase, dNTPs, Mg++, and an inert green tracking dye for easy and direct loading of PCR products onto gels. The broad-use buffer requires only the addition of primers and DNA template for robust amplification, regardless of GC content.
Catalog No. 50-003-32609 Supplier New England Biolabs, Inc. Supplier No. M0580L
The Q5 Quick-Load Hot Start High-Fidelity 2X Master Mix offers high-fidelity performance in a convenient master mix format. The master mix features Q5, a high-fidelity, thermostable DNA polymerase with 3' to 5' exonuclease activity fused to a processivity-enhancing Sso7d domain. With an error rate ~280-fold lower than that of Taq DNA Polymerase, Q5 offers highly accurate amplification and can be used to amplify difficult or long targets (up to 20 kb for simple templates). The addition of a hot start aptamer inhibits polymerase activity at room temperature. The master mix includes hot start polymerase, dNTPs, Mg++, and an inert green tracking dye for easy and direct loading of PCR products onto gels. The broad-use buffer requires only the addition of primers and DNA template for robust amplification, regardless of GC content.