Learn More
New England Biolabs, Inc. Q5® Site-Directed Mutagenesis Kit – 10 reactions
SDP
Supplier:  New England Biolabs, Inc. E0554S
Encompass
The Q5 Site-Directed Mutagenesis Kit enables rapid, site-specific mutagenesis of double-stranded plasmid DNA in less than 2 hours. The kit utilizes the robust Q5 Hot Start High-Fidelity DNA Polymerase along with custom mutagenic primers to create insertions, deletions and substitutions in a wide variety of plasmids. After PCR, the amplified material is added directly to a unique Kinase-Ligase-DpnI (KLD) enzyme mix for rapid (5 minutes), room temperature circularization and template removal. Transformation into high-efficiency chemically-competent E. coli, ensures robust results with plasmids up to at least 20 kb in length.
  • Non-overlapping primer design ensures robust, exponential amplification
  • Intramolecular ligation/transformation into NEB high efficiency competent cells results in high colony yield
  • DpnI background reduction permits a wide range of starting template concentrations
Catalog No. NC0475889