Learn More
New England Biolabs, Inc. Q5U™ Hot Start High-Fidelity DNA Polymerase – 500 units
SDP
Catalog No. NC1737567
Encompass
Q5U Hot Start High-Fidelity DNA Polymerase is a modified version of Q5 High-Fidelity DNA Polymerase, a novel thermostable DNA polymerase that possesses 3' to 5' exonuclease activity, and is fused to a processivity-enhancing Sso7d domain. Q5U contains a mutation in the uracil-binding pocket that enables the ability to read and amplify templates containing uracil and inosine bases.
Catalog No. NC1737567 Supplier New England Biolabs, Inc. Supplier No. M0515L
Q5U Hot Start High-Fidelity DNA Polymerase is a modified version of Q5 High-Fidelity DNA Polymerase, a novel thermostable DNA polymerase that possesses 3' to 5' exonuclease activity, and is fused to a processivity-enhancing Sso7d domain. Q5U contains a mutation in the uracil-binding pocket that enables the ability to read and amplify templates containing uracil and inosine bases.