Learn More
Cayman Chemical Q94 (hydrochloride hydrate), Purity: ≥98%, 10 mg
SDP
Catalog No. 502950800
Encompass_Preferred
A Gαq signaling-biased PAR1 antagonist (IC50 = 916 nM); inhibits thrombin- or a PAR1-activating peptide-induced increases in intracellular calcium mobilization in HMEC-1 cells at 1 and 10 μM; decreases albuminuria in a doxorubicin-induced mouse model of nephropathy at 5 mg/kg per day
Catalog No. 50-295-0800 Supplier Cayman Chemical Supplier No. 3285710 MG
A Gαq signaling-biased PAR1 antagonist (IC50 = 916 nM); inhibits thrombin- or a PAR1-activating peptide-induced increases in intracellular calcium mobilization in HMEC-1 cells at 1 and 10 μM; decreases albuminuria in a doxorubicin-induced mouse model of nephropathy at 5 mg/kg per day