Description
- Ideal for mobile treatment engineers
- Compact and portable design
- Easy to follow instructions including pictograms
- Complete with all necessary equipment
Specifications
Specifications
| pH Range | 0 to 14 |
| Volume (Metric) | 20 mL |
| No. of Tests | 100 Tests |
| Range | 60 to 2000 mg/L CTAB |
| Type | QAC/Cationics Test |