Learn More
LaMotte™ QAC Individual Test Kits
SDPSCI_ED
Catalog No. S72325 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
50 Tests/Pk.
Catalog No. Quantity
S72325 50 Tests/Pk.
1 options
This item is not returnable. View return policy
Catalog No. S72325 Supplier LaMotte™ Supplier No. 3043DR01
Add to Cart
Add to Cart
This item is not returnable. View return policy
Direct Reading Titrator method allows for precise, convenient QAC testing
  • Tests for Quaternary Ammonium Compounds (QACs), such as Alkyl dimethyl benzyl ammonium chloride
  • Range and sensitivity is 0 to 500 ppm in 10 ppm increments
  • Indicator is bromphenol blue concentrations.
  • For QAC testing in industrial water boiling and cooling, water and wastewater, laundry and sanitation.
Specifications
For Use With (Application) QAC testing in industrial water boiling and cooling, water and wastewater, laundry and sanitation
Includes 15mL QAC indicator solution, 60mL QAC titration solution, 1 test tube, plastic with cap, 1 direct reading titrator, 0 to 500 range, 1 brush, test tube
No. of Tests 50
Quantity 50 Tests/Pk.
Range 10 to 500ppm
Test Method BPB direct reading titrator
Type QAC Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov