Learn More
Hach Company Quaternary Ammonium Compound (QAC) Standard Solution, 100 mg/L, 100 mL
Catalog No. 501995952 Shop All Hach Company Products
Encompass
For accuracy checks in Quaternary Ammonium Compound (QAC) determinations. 100 mg/L as CTAB.
Catalog No. 50-199-5952 Supplier Hach Company Supplier No. 2415342
For accuracy checks in Quaternary Ammonium Compound (QAC) determinations. 100 mg/L as CTAB.