Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ QAC Test Tablets

Catalog No. 09053201 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-053-201 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-201 Supplier Lovibond™ Supplier No. 515410BT
Add to Cart
Add to Cart

The QAC Test Tablets are the refill reagent for the Lovibond AF417 MINKIT for detecting Quaternary Ammonium Compounds.

  • Highest accuracy
  • Easy handling
  • Precise dosing
  • Enormous durability

Specifications

Analytes Quaternary ammonium compounds (QACs)
Format Tablet Reagent
For Use With (Application) A titrimetric method for the determination of QAC levels (0-500 mg/L) in detergent solutions. It can also be used as a simple control test for QAC levels at 200 mg/L.
For Use With (Equipment) Lovibond QAC MINIKIT (414170)
No. of Tests Variable
No. per Pack 100 Pk.
Quantity 1 Ea.
Sample Size 50 mL
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.