Learn More
Medchemexpress LLC QAQ dichloride | 98.68% | 567.59 | 25 MG
SDP
Catalog No. 5000345914
Encompass_Preferred
QAQ dichloride is a photoswitchable voltage-gated Nav and Kv channels blocker. It blocks channels when in its trans form (of the azobenzene photoswitch) but not in its cis form. This compound is membrane-impermeant and selectively infiltrates pain-sensing neurons that express endogenous import channels. QAQ dichloride acts as a light-sensitive analgesic and is useful for studying signaling mechanisms in acute and chronic pain.
  • Photoswitchable voltage-gated Nav and Kv channels blocker
  • Blocks channels in trans form, not in cis form
  • Membrane-impermeant
  • Infiltrates pain-sensing neurons
  • Acts as a light-sensitive analgesic
  • Can be used for studying signaling mechanisms in acute and chronic pain
Catalog No. 50-003-45914 Supplier Medchemexpress LLC Supplier No. 25MGHY110358
QAQ dichloride is a photoswitchable voltage-gated Nav and Kv channels blocker. It blocks channels when in its trans form (of the azobenzene photoswitch) but not in its cis form. This compound is membrane-impermeant and selectively infiltrates pain-sensing neurons that express endogenous import channels. QAQ dichloride acts as a light-sensitive analgesic and is useful for studying signaling mechanisms in acute and chronic pain.
  • Photoswitchable voltage-gated Nav and Kv channels blocker
  • Blocks channels in trans form, not in cis form
  • Membrane-impermeant
  • Infiltrates pain-sensing neurons
  • Acts as a light-sensitive analgesic
  • Can be used for studying signaling mechanisms in acute and chronic pain