Learn More
Medchemexpress LLC QAQ dichloride | 99.3% | 567.59 g/mol | C28H44Cl2N6O2 | 5 MG
SDP
Catalog No. 5000263498
Encompass_Preferred
QAQ dichloride is a photoswitchable azobenzene-based open-channel blocker that inhibits voltage-gated sodium and potassium channels in its trans configuration. It is membrane-impermeant and selectively enters neurons that express endogenous import channels, making it a versatile light-sensitive probe for studying neuronal signaling and pain mechanisms in research settings.
  • Photoswitchable control of Nav and Kv channel activity.
  • Reversible channel block via photoisomerization.
  • Selective entry into neurons expressing import channels.
  • Applicable to acute and chronic pain signaling studies.
  • Stable dichloride salt form with defined storage recommendations.
Catalog No. 50-002-63498 Supplier Medchemexpress LLC Supplier No. HY1103585MG
QAQ dichloride is a photoswitchable azobenzene-based open-channel blocker that inhibits voltage-gated sodium and potassium channels in its trans configuration. It is membrane-impermeant and selectively enters neurons that express endogenous import channels, making it a versatile light-sensitive probe for studying neuronal signaling and pain mechanisms in research settings.
  • Photoswitchable control of Nav and Kv channel activity.
  • Reversible channel block via photoisomerization.
  • Selective entry into neurons expressing import channels.
  • Applicable to acute and chronic pain signaling studies.
  • Stable dichloride salt form with defined storage recommendations.