Learn More
Teknova QB Citrate Buffer, pH 3.0. 1000mL, Sterile.
SDP
Catalog No. 50842999
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
This item is not returnable. View return policy
Catalog No. 50-842-999 Supplier Teknova Supplier No. Q2445
Add to Cart
Add to Cart
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.