Learn More
Teknova QB Citrate Buffer, pH 6.0. 1000mL, Sterile
SDP
Catalog No. 502388108
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-8108 Supplier Teknova Supplier No. Q2446
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.