Learn More
Teknova QB Citrate Buffer Solution, pH 5.0. 1000mL, Sterile. 2 Pack.
SDP
Catalog No. 502387656
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7656 Supplier Teknova Supplier No. Q24432PK
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.