Learn More
QBC Diagnostics Autoread™ Plus Dry Centrifugal Hematology System
Catalog No. 23900110 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Type:
Autoread Plus Analyzer
Autoread Plus Analyzer w/ Accessories
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-900-110 Autoread Plus Analyzer w/ Accessories
23-900-109 Autoread Plus Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-900-110 Supplier QBC Diagnostics Supplier No. 429577
Autoread™ Plus Dry Centrifugal Hematology System for cost-effective and maintenance free hematology.
  • Employs dry hematology technology using tubes that are internally coated with necessary reagents—no need for costly liquid reagents.
  • AccuTubes can be used for both venous and capillary blood samples.
  • Yields quantitative values from venous or capillary blood sample for nine parameters:
    • Hematocrit (HCT)
    • Hemoglobin (HgB)
    • Mean Corpuscular Hemoglobin Concentration (MCHC)
    • White Blood Cell Count (WBC)
    • Granulocyte Count/Percentage
    • Lymphocyte and Monocyte Count/Percentage
  • No routine maintenance or calibration necessary—a simple calibration check is run daily on a calibration rod to verify performance.
    • Liquid controls available separately for additional performance monitoring.
  • Optical Signature Analysis automatically scans collection tube eight times, eliminating subjective interpretation.
  • Printouts can include age-and sex-specific normal ranges and error messages to alert user to improper tube preparation, unusual sample results, or other irregular conditions.
  • Analyzer software contains hematology diagnostic reminders (HDR)
  • Users may select from several printed formats for test results:
    • Test values only
    • Test values plus abbreviated HDR comments
    • Test values plus expanded HDR comments
Compliance
Complies with CLIA EQC Option 2.
Specifications
Type Autoread Plus Analyzer w/ Accessories
For Use With (Application) Dry hematology technology using tubes that are internally coated with necessary reagents
For Use With (Equipment) Autoread plus hematology system
Certifications/Compliance CLIA EQC Option 2
Includes Software Cartridge, Workstation, Accutube Starter Kit, Power Pack, and Line Cords for 120/230 V, 50/60 Hz Operation
Electrical Requirements 120/230V, 50/60Hz
Frequency 50/60 Hz
Voltage 120/230 V