Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QBC Diagnostics Blade Lancets

Catalog No. 23900139 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Depth (Metric):
1.8 mm
2.3 mm
For Use With (Application):
Designed for use on adult patients
For pediatric use, Ideal for drawing capillary blood for use with QBC tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (Metric) For Use With (Application)
23-900-139 2.3 mm Designed for use on adult patients
23-900-138 1.8 mm For pediatric use, Ideal for drawing capillary blood for use with QBC tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-139 Supplier QBC Diagnostics Supplier No. 420001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These spring loaded Blade Lancets are ideal for drawing blood. For use with the STAR and Autoread Plus Tubes.

  • For use with Autoread Plus Dry Hematology Analyzer.

Specifications

Product Type Blade Lancet
Certifications/Compliance CLIA EQC Option 2
Depth (English) 0.09 in.
Depth (Metric) 2.3 mm
Quantity 100 Pk.
Color Blue
For Use With (Equipment) Autoread Plus Hematology System
For Use With (Application) Designed for use on adult patients
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.