Learn More
QBC Diagnostics Capillary Tubes
Catalog No. 23900100 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Quantity:
100 Pk.
Catalog No. Quantity
23-900-100 100 Pk.
1 options
Catalog No. 23-900-100 Supplier QBC Diagnostics Supplier No. 424241
Add to Cart
Add to Cart
Capillary Tubes for use with Autoread Plus Dry Hematology Analyzer.
  • For use with Autoread Plus Dry Hematology Analyzer.
Specifications
Product Type Capillary Tube
For Use With (Equipment) Autoread Plus Hematology System
Quantity 100 Pk.
Includes Individual Trays of 10 Tubes with External Floats and Manual Closures
For Use With (Application) Can be used with Blood samples taken from a finger or heel stick
Certifications/Compliance CLIA EQC Option 2
Volume (Metric) 60 μL