Learn More
QBC Diagnostics Venous Tubes
Catalog No. 23900104 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Quantity:
100 Pk.
Catalog No. Quantity
23-900-104 100 Pk.
1 options
Catalog No. 23-900-104 Supplier QBC Diagnostics Supplier No. 424240
Add to Cart
Add to Cart
Venous Tubes for hematology determinations with Venous blood using the Autoread Analyzer.
  • For use with Autoread Plus Dry Hematology Analyzer.
  • For In Vitro Diagnostics use.
Specifications
Product Type Blood Collection Tube
For Use With (Equipment) Autoread Plus Hematology System
Quantity 100 Pk.
Includes Individual Trays of 10 Tubes with External Floats and Manual Closures
For Use With (Application) Can be used with the pipetter to draw samples from venous tubes
Certifications/Compliance CLIA EQC Option 2
Volume (Metric) 111 μL