Learn More
QBC Diagnostics Capillary Centrifuge
Catalog No. 23900111 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Capacity:
60 μL
Catalog No. Capacity
23-900-111 60 μL
1 options
Catalog No. 23-900-111 Supplier QBC Diagnostics Supplier No. 425740
Add to Cart
Add to Cart
The Capillary Centrifuge is an ultra-compact centrifuge designed for centrifuging tubes of patient blood samples for use in the Autoread Plus or Malaria Test Systems.
  • In as little as five (5) minutes, the Capillary Centrifuge can centrifuge up to twenty (20) samples at one time.
Specifications
Capacity 60 μL
Certifications/Compliance CLIA EQC Option 2
Includes Power Pack and U.S. and European Line Cords for 120/230 V, 50/60 Hz Operation
For Use With (Application) Used to spin all QBC tubes (except for the STAR Tube) for testing in the autoread plus analyzer
Product Type Capillary Centrifuge