Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QBC Diagnostics F.A.S.T. Prepackaged AFB Kits

Catalog No. 23900145 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Type:
F.A.S.T AFB Smear Stain Kit
F.A.S.T. AFB Smear Kit with Digestion Solution
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-900-145 F.A.S.T. AFB Smear Kit with Digestion Solution
23-900-144 F.A.S.T AFB Smear Stain Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-900-145 Supplier QBC Diagnostics Supplier No. 427408

Includes

QBC F.A.S.T AFB Smear Kit

  • F.A.S.T Auramine O Stain Kit (120mL)
  • Quality control slides (5)
  • F.A.S.T SureFocus Miscroscope Slides (432)
  • F.A.S.T Sputum Cups (400)
  • F.A.S.T Applicator Sticks (400)
QBC F.A.S.T AFB Smear Kit with Digestion Solution
  • F.A.S.T Auramine O Stain Kit (120mL)
  • Quality control slides (5)
  • F.A.S.T SureFocus Miscroscope Slides (432)
  • Bottles of F.A.S.T Sputum Digestion Solution (27)

Complete AFB staining systems

  • Three minute staining
  • Supplies for 400 tests

Specifications

Type F.A.S.T. AFB Smear Kit with Digestion Solution
Quantity 400 Tests
Includes 120mL Auramine O stain kit, 5 quality control slides, 432 SureFocus miscroscope slides, 27 sputum digestion solution
For Use With (Application) Detect positive AFB (tuberculosis) sputum samples
Storage Requirements 15° to 25°C
Stain Type Auramine O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.