Learn More
QBC Diagnostics ParaLens™ Advance Microscope System: Batteries
Catalog No. 23900205 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Product Type:
12V Battery clip to lighter adapter
Replacement solar cell battery pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-900-205 12V Battery clip to lighter adapter
23-900-201 Replacement solar cell battery pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-900-205 Supplier QBC Diagnostics Supplier No. 427721
Specifications
Includes Positive and negative terminal clips and a 12V lighter socket that can be used with the lighter adapter to light source cable
For Use With (Application) Power the ParaLens using a 12V battery
For Use With (Equipment) ParaLens advance microscope attachment LED light source
Product Type 12V Battery clip to lighter adapter