Learn More
QBC Diagnostics ParaLens™ Advance Microscope System: Portability Pack
Catalog No. 23900200 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Product Type:
Portability pack
Catalog No. Product Type
23-900-200 Portability pack
1 options
Catalog No. 23-900-200 Supplier QBC Diagnostics Supplier No. 424340
Specifications
Includes Solar cell battery pack, solar cell wall charger (for use with 100/240V operation), USB to 3.5mm light source cable, 12V battery clip to lighter adapter, lighter adapter to 3.5mm light source cable and complete fluorescence system case
For Use With (Application) Power the ParaLens advance microscope attachment LED light source in remote locations
For Use With (Equipment) ParaLens advance microscope attachment LED light source
Product Type Portability pack