Description
- Detect positive AFB (Tuberculosis) sputum samples in as little as three minutes
- Multiple kit configurations are available to suit the requirements of any laboratory
Specifications
Specifications
| Includes | Research grade microscope with 4X, 10X, 40X and 100X objectives, ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon™ 755VES centrifuge (230V, 50Hz) and QBC F.A.S.T. AFB smear Kit with digestion solution |
| Magnification Power | 4X, 10X, 40X, 100X |
| For Use With (Application) | AFB staining and review |