Learn More
QBC Diagnostics F.A.S.T™. AFB System
Catalog No. 23900142 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Magnification Power:
20X, 40X
4X, 10X, 40X, 100X
Includes:
ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon 755VES centrifuge (115V, 60Hz) and F.A.S.T. AFB smear kit with digestion solution
ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon 755VES centrifuge (230V, 50Hz) and F.A.S.T. AFB smear kit with digestion solution
Research grade microscope with 4X, 10X, 40X and 100X objectives, ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon™ 755VES centrifuge (115V, 60Hz) and QBC F.A.S.T. AFB smear Kit with digestion solution
Research grade microscope with 4X, 10X, 40X and 100X objectives, ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon™ 755VES centrifuge (230V, 50Hz) and QBC F.A.S.T. AFB smear Kit with digestion solution
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power Includes
23-900-142 20X, 40X ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon 755VES centrifuge (115V, 60Hz) and F.A.S.T. AFB smear kit with digestion solution
23-900-140 4X, 10X, 40X, 100X Research grade microscope with 4X, 10X, 40X and 100X objectives, ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon™ 755VES centrifuge (115V, 60Hz) and QBC F.A.S.T. AFB smear Kit with digestion solution
23-900-143 20X, 40X ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon 755VES centrifuge (230V, 50Hz) and F.A.S.T. AFB smear kit with digestion solution
23-900-141 4X, 10X, 40X, 100X Research grade microscope with 4X, 10X, 40X and 100X objectives, ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon™ 755VES centrifuge (230V, 50Hz) and QBC F.A.S.T. AFB smear Kit with digestion solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-900-142 Supplier QBC Diagnostics Supplier No. 427420
AFB (Tuberculosis) detection in just 3 minutes
  • Detect positive AFB (Tuberculosis) sputum samples in as little as three minutes
  • Multiple kit configurations are available to suit the requirements of any laboratory
Specifications
Includes ParaLens advance microscope attachment with 20X and 40X objectives, QBC Horizon 755VES centrifuge (115V, 60Hz) and F.A.S.T. AFB smear kit with digestion solution
Magnification Power 20X, 40X
For Use With (Application) AFB staining and review