Learn More
QBC Diagnostics Malaria Test
Catalog No. 23900134 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Quantity:
1 Ea.
100 Tests
2000 Tests
Type:
Malaria Test
Test System with ParaLens Advance and Centrifuge
Test System with ParaLens Advance, Microscope, and Centrifuge
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-900-134 2000 Tests Malaria Test
23-900-133 100 Tests Malaria Test
23-900-131 1 Ea. Test System with ParaLens Advance, Microscope, and Centrifuge
23-900-132 1 Ea. Test System with ParaLens Advance and Centrifuge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-900-134 Supplier QBC Diagnostics Supplier No. 253005
Add to Cart
Add to Cart
Malaria Test for proven malaria detection.
  • The QBC Malaria Test is a fluorescence microscopy-based malaria diagnostic test embraced by users around the world.
  • More sensitive than Giemsa thick films, it can detect as little as one parasite per μL of blood and establish diagnosis earlier than thick film in 47% of low parasitemia (<10 parasites per μL) cases.
QBC Malaria Test Features:
  • Test consists of a capillary tube coated with anticoagulants and acridine orange used to stain malaria parasites for detection using fluorescence microscopy.
  • Results are possible in just 8 minutes, a fraction of the time of thick film methods.
  • Training time is reduced, as users with just five days of training can produce results comparable to an experienced microscopist using thick film.
  • Simple set-up and review: special QBC technology uses a single capillary tube to collect, stain and process blood samples.
  • Principles of centrifugation and fluorescence improve review by concentrating easily visible parasites in specific areas of the tube.
  • QBC Malaria Test was the first malaria diagnostic test to receive FDA clearance.
  • QBC Malaria Test is internally coated with fluorescent acridine orange stain to improve the visibility of malaria parasites; when excited by blue light (∽460 nm), parasites will fluoresce brightly against a dark background, as seen in the micrographs.
  • Concentrates 50 μL blood samples, allowing users to quickly and easily see a far greater number of parasites than Giemsa tests, which use just 2 μL of blood; provides unmatched results in cases of low parasitemia.
  • ParaLens Advance attachment provides fluorescence capabilities to any light microscope through the use of a durable, long lasting LED light source; with simple setup and multiple power options, it can be transported wherever testing is most convenient.
QBC Malaria Test System with ParaLens Advance™, Microscope, and Centrifuge
  • A complete transmission light microscope and epi-fluorescence microscopy system for malaria parasite staining and review.
  • Includes research grade microscope with 4X, 10X, 40X and 100X objectives, ParaLens Advance microscope attachment with 60X objective, Capillary Centrifuge (120/230V, 50/60 Hz operation), and Malaria Test (box of 2000).
QBC Malaria Test System with ParaLens Advance and Centrifuge
  • An epi-fluorescence microscopy system for malaria parasite staining and review.
  • Includes ParaLens Advance microscope attachment with 60X objective, Capillary Centrifuge (120/230 V, 50/60 Hz operation), and Malaria Test (box of 2000).
Test Kits:
  • QBC Malaria Test (Box of 100): Includes 10 individual trays of 10 tests, with precision plastic floats and closures.
  • QBC Malaria Test (Box of 2000): Includes 8 individual boxes of 250 tests, with precision plastic floats and closures.
Accessories:
  • 1.8 mm Blade Lancets: spring loaded lancets, ideal for drawing capillary blood for use with QBC Tubes, for pediatric use.
  • 2.3 mm Blade Lancets: spring loaded lancets, for use on adult patients.
Compliance
U.S. FDA cleared
Specifications
Detectable Analytes Malaria Parasites
Type Malaria Test
Includes 8 Individual Boxes of 250 Tests, with Precision Plastic Floats and Closures
Quantity 2000 Tests