Learn More
QBC Diagnostics Star™ Dry Centrifugal Hematology System
Catalog No. 23900103 Shop All Drucker Diagnostics Products
Change view
Click to view available options
For Use With (Equipment):
QBC Star Hematology Analyzer
Catalog No. For Use With (Equipment)
23-900-103 QBC Star Hematology Analyzer
1 options
Catalog No. 23-900-103 Supplier QBC Diagnostics Supplier No. 429625
Add to Cart
Add to Cart
Star™ Dry Centrifugal Hematology System is a compact and easy to use complete blood count analyzer designed to bring CBCs to the point of care.
  • Compact and easy to use complete blood count analyzer designed to bring CBCs to the point of care.
  • Designed for labs performing up to 20 CBC tests daily.
  • Employs dry hematology technology using patented STAR tubes that are internally coated with necessary reagents — no need for costly liquid reagents.
  • STAR tubes can be used for both venous and capillary blood samples.
  • Yields quantitative values from venous or capillary blood sample for nine parameters:
    • Hematocrit (HCT)
    • Hemoglobin (HgB)
    • Mean Corpuscular Hemoglobin Concentration (MCHC)
    • White Blood Cell Count (WBC)
    • Granulocyte Count/Percentage
    • Lymphocyte and Monocyte Count/Percentage
  • Self-contained instrument includes centrifuge, analyzer and printer in a small 16 × 16 in. footprint.
  • Fully automated with easy operation — simply add a blood sample to the tube, place it in the system, press START and wait for the results in minutes.
  • Optical Signature Analysis automatically scans each tube eight times for increased precision.
  • No routine maintenance, calibration or trained specialist required.
Compliance
Pending CLIA waiver. Complies with CLIA EQC Option 1.
Specifications
Type QBC Star Tube
For Use With (Equipment) QBC Star Hematology Analyzer
Sample Size 70 μL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov