Learn More
Quality Biological Inc QBSF-60, 500 mL
SDP
Catalog No. NC0823508
Encompass_Preferred
QBSF-60 is a proprietary serum-free medium ¿ Supports the expansion of human hematopoietic stem/progenitor cells. ¿ Supports CD34+ cells derived from normal bone marrow, cord blood, and peripheral blood stem cells mobilized with cytokines for ex-vivo expansion and differentiation to erythrocytes, granulocytes, and lymphocytes. ¿ Storage Conditions: 2¿C to 8¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ pH: 7.0¿ 0.2 ¿ Osmolality: 285 ¿ 3% mOsm/Kg H20 ¿ Endotoxin, mycoplasma, and performance tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ Max Shelf Life: 24 months
Catalog No. NC0823508 Supplier Quality Biological Inc Supplier No. 160204101
QBSF-60 is a proprietary serum-free medium ¿ Supports the expansion of human hematopoietic stem/progenitor cells. ¿ Supports CD34+ cells derived from normal bone marrow, cord blood, and peripheral blood stem cells mobilized with cytokines for ex-vivo expansion and differentiation to erythrocytes, granulocytes, and lymphocytes. ¿ Storage Conditions: 2¿C to 8¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ pH: 7.0¿ 0.2 ¿ Osmolality: 285 ¿ 3% mOsm/Kg H20 ¿ Endotoxin, mycoplasma, and performance tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ Max Shelf Life: 24 months