Learn More
Bioworld QBT Buffer, 500 mL
Catalog No. 50196887
Encompass_Preferred
Product Description QBT buffer is a plasmid DNA purification buffer. It is composed of 750 mM NaCl, 50 mM MOPS, 15% isopropanol, 0.15% Triton X-100 and has a pH value of 7.0.
Catalog No. 50-196-887 Supplier Bioworld Supplier No. 107600352
Product Description QBT buffer is a plasmid DNA purification buffer. It is composed of 750 mM NaCl, 50 mM MOPS, 15% isopropanol, 0.15% Triton X-100 and has a pH value of 7.0.