Learn More
Bioworld QBT Buffer, 250 mL
Catalog No. 50196886
Encompass_Preferred
Product Description QBT buffer is a plasmid DNA purification buffer. It is composed of 750 mM NaCl, 50 mM MOPS, 15% isopropanol, 0.15% Triton X-100 and has a pH value of 7.0.
Catalog No. 50-196-886 Supplier Bioworld Supplier No. 107600351
Product Description QBT buffer is a plasmid DNA purification buffer. It is composed of 750 mM NaCl, 50 mM MOPS, 15% isopropanol, 0.15% Triton X-100 and has a pH value of 7.0.