Learn More
Molecular Devices Molecular Devices FLIPR Calcium 4 Kit, No-Wash, Fluorescence, 10 Plates
Catalog No. 5000014405
Encompass_Preferred
Molecular Devices FLIPR Calcium 4 Assay Kit Explorer provides a no-wash, fluorescence-based method for detecting intracellular calcium changes with improved signal-to-noise using a novel masking dye.
  • Designed for no-wash fluorescence calcium assays.
  • Includes enough reagent for 10 plates in the Explorer configuration.
  • Optimized for small calcium responses with reduced background.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Supplied as reagent vials and a dilution/assay buffer to prepare plates.
Catalog No. 50-000-14405 Supplier Molecular Devices Supplier No. R8142
Molecular Devices FLIPR Calcium 4 Assay Kit Explorer provides a no-wash, fluorescence-based method for detecting intracellular calcium changes with improved signal-to-noise using a novel masking dye.
  • Designed for no-wash fluorescence calcium assays.
  • Includes enough reagent for 10 plates in the Explorer configuration.
  • Optimized for small calcium responses with reduced background.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Supplied as reagent vials and a dilution/assay buffer to prepare plates.