Learn More
Molecular Devices Molecular Devices QBT Fatty Acid Uptake Assay, Bulk, BODIPY-dodecanoic, 96/384-well
Catalog No. 5000014435
Encompass_Preferred
The QBT Fatty Acid Uptake Assay Kit is a homogeneous, fluorescence-based assay for measuring live-cell fatty acid uptake kinetics and screening fatty acid transporters in high-throughput formats. The assay uses a fluorescent fatty acid analog with a quench reagent to reduce extracellular background and enable kinetic readings.
  • Homogeneous fluorescence assay suitable for high-throughput screening.
  • Uses a BODIPY-dodecanoic fluorescent fatty acid analog and quench reagent.
  • Bulk kit contains 10 vials, sufficient for approximately 50 plates.
  • Compatible with 96-well and 384-well microplate formats.
  • Measure kinetics with bottom-read fluorescence, excitation ~470-500 nm, emission ~500-560 nm.
  • Store kit at -20°C; formulated loading buffer stable up to 8 hours at room temperature.
Catalog No. 50-000-14435 Supplier Molecular Devices Supplier No. R8133
The QBT Fatty Acid Uptake Assay Kit is a homogeneous, fluorescence-based assay for measuring live-cell fatty acid uptake kinetics and screening fatty acid transporters in high-throughput formats. The assay uses a fluorescent fatty acid analog with a quench reagent to reduce extracellular background and enable kinetic readings.
  • Homogeneous fluorescence assay suitable for high-throughput screening.
  • Uses a BODIPY-dodecanoic fluorescent fatty acid analog and quench reagent.
  • Bulk kit contains 10 vials, sufficient for approximately 50 plates.
  • Compatible with 96-well and 384-well microplate formats.
  • Measure kinetics with bottom-read fluorescence, excitation ~470-500 nm, emission ~500-560 nm.
  • Store kit at -20°C; formulated loading buffer stable up to 8 hours at room temperature.